Fmoc-β-methyl-DL-phenylalanine
Supplier: Matrix Scientific
041996-100MG
041996-1G
101854-030EA
138.46
USD
101854-030
101854-032
Fmoc-β-methyl-DL-phenylalanine
Fmoc-beta-methyl-DL-phenylalanine
Hazard: Irritant
Formula:
C₂₅H₂₃NO₄ MW: 401.47 g/mol |
MDL Number:
MFCD01823266 CAS Number: 1214028-21-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...