Fmoc-α-methyl-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methyl-3-phenylpropanoic acid, Fmoc-α-Me-Phe-OH

041995-250MG 041995-1G
101854-026EA 386.15 USD
101854-026 101854-028
Fmoc-α-methyl-L-phenylalanine
Fmoc-alpha-methyl-L-phenylalanine

CAS: 135944-05-7 MDL Number: MFCD02682285 Molecular Formula: C25H23NO4 Molecular Weight: 401,47 Hazard: Irritant

Formula: C₂₅H₂₃NO₄
MW: 401.47 g/mol
MDL Number: MFCD02682285
CAS Number: 135944-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR