Fmoc-α-methyl-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: (R)-N-FMOC-α-Methylphenylalanine, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methyl-3-phenylpropanoic acid, Fmoc-α-Me-D-Phe-OH

041994-250MG 041994-1G 041994-5G
101854-020EA 386.15 USD
101854-020 101854-022 101854-024
Fmoc-α-methyl-D-phenylalanine
Fmoc-alpha-methyl-D-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₃NO₄
MDL Number: MFCD02259493
CAS Number: 152436-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR