Fmoc-α-methyl-3-methoxy-DL-phenylalanine

Supplier: Matrix Scientific

041993-100MG 041993-500MG
101854-016EA 275.38 USD
101854-016 101854-018
Fmoc-α-methyl-3-methoxy-DL-phenylalanine
Fmoc-alpha-methyl-3-methoxy-DL-phenylalanine

Hazard: Irritant

Formula: C₂₆H₂₅NO₅
MW: 431.49 g/mol
MDL Number: MFCD02682277
CAS Number: 1185301-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR