Fmoc-hydroxy-Tic-OH

Supplier: Matrix Scientific

041992-250MG 041992-1G 041992-5G
101854-010EA 249.23 USD
101854-010 101854-012 101854-014
Fmoc-hydroxy-Tic-OH
Fmoc-hydroxy-Tic-OH

Hazard: Irritant

Formula: C₂₅H₂₁NO₅
MDL Number: MFCD00792388
CAS Number: 178432-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR