Fmoc-5-bromo-2-methoxy-L-phenylalanine

Supplier: Matrix Scientific

041990-500MG 041990-1G
101854-004EA 598.46 USD
101854-004 101854-006
Fmoc-5-bromo-2-methoxy-L-phenylalanine
Fmoc-5-bromo-2-methoxy-L-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₂BrNO₅
MDL Number: MFCD01311746
CAS Number: 220497-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR