Fmoc-5-bromo-2-methoxy-D-phenylalanine

Supplier: Matrix Scientific

041989-500MG 041989-1G
101854-000EA 686.15 USD
101854-000 101854-002
Fmoc-5-bromo-2-methoxy-D-phenylalanine
Fmoc-5-bromo-2-methoxy-D-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₂BrNO₅
MW: 496.37 g/mol
MDL Number: MFCD01311773
CAS Number: 220497-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR