Fmoc-4-nitro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-p-nitro-D-Phe-OH, Fmoc-D-Phe(4-NO₂)-OH, Fmoc-D-Phe(4-NO2)-OH

041987-1G 041987-5G
101853-992EA 224.62 USD
101853-992 101853-994
Fmoc-4-nitro-D-phenylalanine
Fmoc-4-nitro-D-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀N₂O₆
MW: 432.432 g/mol
MDL Number: MFCD00237029
CAS Number: 177966-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR