Fmoc-L-4-methylphe

Supplier: Matrix Scientific

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methyl-L-phenylalanine, N-Fmoc-4-methyl-L-phenylalanine, Fmoc-Phe(4-Me)-OH, Fmoc-p-Me-Phe-OH

041986-1G 041986-5G
101853-988EA 300 USD
101853-988 101853-990
Fmoc-L-4-methylphe
Fmoc-L-4-methylphe

Hazard: Irritant

Formula: C₂₅H₂₃NO₄
MW: 401.462 g/mol
MDL Number: MFCD00270197
CAS Number: 199006-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR