Fmoc-4-methyl-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-p-Me-D-Phe-OH, Fmoc-D-Phe(4-Me)-OH

041985-1G 041985-5G
101853-984EA 300 USD
101853-984 101853-986
Fmoc-4-methyl-D-phenylalanine
Fmoc-4-methyl-D-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₃NO₄
MW: 401.462 g/mol
MDL Number: MFCD00270196
CAS Number: 204260-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR