Fmoc-4-iodo-L-phenylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: Fmoc-p-iodo-Phe-OH, Fmoc-Phe(4-I)-OH

041984-1G 041984-5G
101853-980EA 106.15 USD
101853-980 101853-982
Fmoc-4-iodo-L-phenylalanine ≥95%
Fmoc-4-iodo-L-phenylalanine

Hazard: Irritant/Light Sensitive

Formula: C₂₄H₂₀INO₄
MW: 513.331 g/mol
Melting Pt: 213…217 °C
MDL Number: MFCD00672340
CAS Number: 82565-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR