N-Fmoc-4-iodo-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Phe(4-I)-OH

041983-1G 041983-5G
101853-976EA 187.69 USD
101853-976 101853-978
N-Fmoc-4-iodo-D-phenylalanine
N-Fmoc-4-iodo-D-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀INO₄
MW: 513.331 g/mol
MDL Number: MFCD00672557
CAS Number: 205526-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR