Fmoc-Phe(4-CN)-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-4-cyano-Phe-OH

041982-1G 041982-5G
101853-972EA 547.69 USD
101853-972 101853-974
Fmoc-Phe(4-CN)-OH
Fmoc-Phe(4-CN)-OH

Hazard: Irritant

Formula: C₂₅H₂₀N₂O₄
MW: 412.445 g/mol
MDL Number: MFCD00672559
CAS Number: 173963-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR