Fmoc-4-cyano-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Phe(4-CN)-OH

041981-1G 041981-5G
101853-968EA 187.69 USD
101853-968 101853-970
Fmoc-4-cyano-D-phenylalanine
Fmoc-4-cyano-D-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₀N₂O₄
MW: 412.45 g/mol
MDL Number: MFCD00672560
CAS Number: 205526-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR