Fmoc-4-chloro-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-4-chloro-Phe-OH, N-Fmoc-4-chloro-L-phenylalanine, Fmoc-Phe(4-Cl)-OH

041980-1G 041980-5G
101853-964EA 138.46 USD
101853-964 101853-966
Fmoc-4-chloro-L-phenylalanine
Fmoc-4-chloro-L-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀ClNO₄
MW: 421.877 g/mol
MDL Number: MFCD00080281
CAS Number: 175453-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR