Fmoc-4-chloro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: N-Fmoc-4-chloro-D-phenylalanine, Fmoc-D-Phe(4-Cl)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-chlorophenyl)propanoic acid, Fmoc-p-chloro-D-Phe-OH

041979-1G 041979-5G
101853-960EA 138.46 USD
101853-960 101853-962
Fmoc-4-chloro-D-phenylalanine
Fmoc-4-chloro-D-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀ClNO₄
MW: 421.877 g/mol
MDL Number: MFCD00077066
CAS Number: 142994-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR