Fmoc-L-Phe(4-Br)-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-Phe(4-Br)-OH

041978-1G 041978-5G
101853-956EA 73.85 USD
101853-956 101853-958
Fmoc-L-Phe(4-Br)-OH
Fmoc-L-Phe(4-Br)-OH

Hazard: Irritant

Formula: C₂₄H₂₀BrNO₄
MW: 466.30 g/mol
MDL Number: MFCD00273460
CAS Number: 198561-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR