Fmoc-D-Bpa-OH

Supplier: Matrix Scientific

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-benzoylphenyl)propanoic acid, Fmoc-p-Bz-D-Phe-OH, N-Fmoc-4-benzoyl-D-phenylalanine

041974-250MG 041974-1G
101853-942EA 224.62 USD
101853-942 101853-944
Fmoc-D-Bpa-OH
Fmoc-D-Bpa-OH

Hazard: Irritant

Formula: C₃₁H₂₅NO₅
MW: 491.543 g/mol
MDL Number: MFCD00237666
CAS Number: 117666-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR