Fmoc-4-amino-L-phenylalanine

Supplier: Matrix Scientific

041973-1G 041973-5G
101853-938EA 164.62 USD
101853-938 101853-940
Fmoc-4-amino-L-phenylalanine
Fmoc-4-amino-L-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₂N₂O₄
MDL Number: MFCD00237664
CAS Number: 95753-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR