Fmoc-4-[2-(boc-amino)ethoxy]-L-phenylalanine
Supplier: Matrix Scientific
Synonyms:
Fmoc-4-[2-(Boc-amino)ethoxy]-L-phenylalanine
041971-250MG
041971-1G
101853-930EA
326.15
USD
101853-930
101853-932
Fmoc-4-[2-(boc-amino)ethoxy]-L-phenylalanine
Fmoc-4-[2-(boc-amino)ethoxy]-L-phenylalanine
Hazard: Irritant
Formula:
C₃₁H₃₄N₂O₇ MW: 546.63 g/mol |
MDL Number:
MFCD07781260 CAS Number: 1013883-02-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...