N-Fmoc-4-(trifluoromethyl)-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-Phe(4-CF3)-OH

041970-1G 041970-5G
101853-926EA 590.77 USD
101853-926 101853-928
N-Fmoc-4-(trifluoromethyl)-L-phenylalanine
N-Fmoc-4-(trifluoromethyl)-L-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₀F₃NO₄
MW: 455.433 g/mol
MDL Number: MFCD00797581
CAS Number: 247113-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR