Fmoc-4-(boc-aminomethyl)-D-phenylalanine

Supplier: Matrix Scientific

041965-1G 041965-5G
101853-906EA 598.46 USD
101853-906 101853-908
Fmoc-4-(boc-aminomethyl)-D-phenylalanine
Fmoc-4-(boc-aminomethyl)-D-phenylalanine

Hazard: Irritant

Formula: C₃₀H₃₂N₂O₆
MW: 516.6 g/mol
MDL Number: MFCD02094578
CAS Number: 268731-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR