Fmoc-4-(boc-amino)-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-Phe(4-NHBoc)-OH, Fmoc-4-(Boc-amino)-L-phenylalanine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-((tert-butoxycarbonyl)amino)phenyl)propanoic acid

041964-1G 041964-5G
101853-902EA 363.08 USD
101853-902 101853-904
Fmoc-4-(boc-amino)-L-phenylalanine
Fmoc-4-(boc-amino)-L-phenylalanine

Hazard: Irritant

Formula: C₂₉H₃₀N₂O₆
MW: 502.57 g/mol
MDL Number: MFCD00237007
CAS Number: 174132-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR