Fmoc-D-Phe(4-NHBoc)-OH
Supplier: Matrix Scientific
Synonyms:
Fmoc-p-amino-D-Phe(Boc)-OH, 4-(Boc-amino)-N-Fmoc-D-phenylalanine
041963-1G
041963-5G
101853-898EA
412.31
USD
101853-898
101853-900
Fmoc-D-Phe(4-NHBoc)-OH
Fmoc-D-Phe(4-NHBoc)-OH
Hazard: Irritant
Formula:
C₂₉H₃₀N₂O₆ MW: 502.567 g/mol |
MDL Number:
MFCD00798642 CAS Number: 214750-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...