Fmoc-3-nitro-L-tyrosine

Supplier: Matrix Scientific

Synonyms: Fmoc-Tyr(3-NO₂)-OH, Fmoc-3-nitro-Tyr-OH, N-Fmoc-3-nitro-L-tyrosine, Fmoc-Tyr(3-NO2)-OH

041962-1G 041962-5G
101853-894EA 124.62 USD
101853-894 101853-896
Fmoc-3-nitro-L-tyrosine
Fmoc-3-nitro-L-tyrosine

Hazard: Irritant

Formula: C₂₄H₂₀N₂O₇
MW: 448.431 g/mol
MDL Number: MFCD00171383
CAS Number: 136590-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR