Fmoc-3-nitro-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-Phe(3-NO2)-OH, Fmoc-Phe(3-NO₂)-OH

041961-1G 041961-5G
101853-890EA 386.15 USD
101853-890 101853-892
Fmoc-3-nitro-L-phenylalanine
Fmoc-3-nitro-L-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀N₂O₆
MW: 432.44 g/mol
MDL Number: MFCD01317718
CAS Number: 206060-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR