N-Fmoc-3-nitro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: 3-Nitro-D-phenylalanine, N-FMOC protected, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-nitrophenyl)propanoic acid

041960-1G 041960-5G
101853-886EA 386.15 USD
101853-886 101853-888
N-Fmoc-3-nitro-D-phenylalanine
N-Fmoc-3-nitro-D-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀N₂O₆
MW: 432.432 g/mol
MDL Number: MFCD01317715
CAS Number: 478183-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR