Fmoc-3-methyl-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: 3-Methyl-L-phenylalanine, N-FMOC protected, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(m-tolyl)propanoic acid, Fmoc-Phe(3-Me)-OH

041959-1G 041959-5G
101853-882EA 300 USD
101853-882 101853-884
Fmoc-3-methyl-L-phenylalanine
Fmoc-3-methyl-L-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₃NO₄
MW: 401.47 g/mol
MDL Number: MFCD01863050
CAS Number: 211637-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR