Fmoc-3-methyl-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Phe(3-Me)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(m-tolyl)propanoic acid

041958-1G 041958-5G
101853-878EA 300 USD
101853-878 101853-880
Fmoc-3-methyl-D-phenylalanine
Fmoc-3-methyl-D-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₃NO₄
MW: 401.47 g/mol
MDL Number: MFCD01863049
CAS Number: 352351-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR