Fmoc-3-iodo-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-Phe(3-I)-OH

041955-1G 041955-5G
101853-870EA 224.62 USD
101853-870 101853-872
Fmoc-3-iodo-L-phenylalanine
Fmoc-3-iodo-L-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀INO₄
MW: 513.34 g/mol
MDL Number: MFCD01632252
CAS Number: 210282-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR