Fmoc-3-iodo-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Phe(3-I)-OH

041954-1G 041954-5G
101853-866EA 224.62 USD
101853-866 101853-868
Fmoc-3-iodo-D-phenylalanine
Fmoc-3-iodo-D-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀INO₄
MW: 513.34 g/mol
MDL Number: MFCD03840402
CAS Number: 478183-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR