Fmoc-3-fluoro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Phe(3-F)-OH

041952-1G 041952-5G
101853-858EA 187.69 USD
101853-858 101853-860
Fmoc-3-fluoro-D-phenylalanine
Fmoc-3-fluoro-D-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀FNO₄
MW: 405.43 g/mol
MDL Number: MFCD00672554
CAS Number: 198545-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR