Fmoc-3-fluoro-DL-tyrosine

Supplier: Matrix Scientific

041951-100MG 041951-1G
101853-854EA 283.08 USD
101853-854 101853-856
Fmoc-3-fluoro-DL-tyrosine
Fmoc-3-fluoro-DL-tyrosine

Hazard: Irritant

Formula: C₂₄H₂₀FNO₅
MDL Number: MFCD02682269
CAS Number: 1219389-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR