Fmoc-3-cyano-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Phe(3-CN)-OH

041949-1G 041949-5G
101853-846EA 547.69 USD
101853-846 101853-848
Fmoc-3-cyano-D-phenylalanine
Fmoc-3-cyano-D-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₀N₂O₄
MW: 412.45 g/mol
MDL Number: MFCD00797586
CAS Number: 205526-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR