Fmoc-3-chloro-L-tyrosine

Supplier: Matrix Scientific

Synonyms: Fmoc-Tyr(3-Cl)-OH

041948-1G 041948-5G
101853-842EA 661.54 USD
101853-842 101853-844
Fmoc-3-chloro-L-tyrosine
Fmoc-3-chloro-L-tyrosine

Hazard: Irritant

Formula: C₂₄H₂₀ClNO₅
MDL Number: MFCD02682261
CAS Number: 478183-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR