3-Chloro-N-fmoc-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-Phe(3-Cl)-OH

041947-1G 041947-5G
101853-838EA 69.23 USD
101853-838 101853-840
3-Chloro-N-fmoc-L-phenylalanine
3-Chloro-N-fmoc-L-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀ClNO₄
MW: 421.877 g/mol
MDL Number: MFCD00672548
CAS Number: 198560-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR