Fmoc-3-chloro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Phe(3-Cl)-OH

041946-1G 041946-5G
101853-834EA 138.46 USD
101853-834 101853-836
Fmoc-3-chloro-D-phenylalanine
Fmoc-3-chloro-D-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀ClNO₄
MW: 421.88 g/mol
MDL Number: MFCD00672549
CAS Number: 205526-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR