Fmoc-3-bromo-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-Phe(3-Br)-OH

041945-500MG 041945-1G
101853-830EA 547.69 USD
101853-830 101853-832
Fmoc-3-bromo-L-phenylalanine
Fmoc-3-bromo-L-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀BrNO₄
MW: 466.34 g/mol
MDL Number: MFCD01311744
CAS Number: 220497-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR