Fmoc-3-bromo-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-bromophenyl)propanoic acid, Fmoc-D-Phe(3-Br)-OH

041944-500MG 041944-1G
101853-826EA 547.69 USD
101853-826 101853-828
Fmoc-3-bromo-D-phenylalanine
Fmoc-3-bromo-D-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀BrNO₄
MW: 466.34 g/mol
MDL Number: MFCD01311771
CAS Number: 220497-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR