Fmoc-3-amino-L-tyrosine

Supplier: Matrix Scientific

041943-1G 041943-5G
101853-822EA 1207.69 USD
101853-822 101853-824
Fmoc-3-amino-L-tyrosine
Fmoc-3-amino-L-tyrosine

Hazard: Irritant

Formula: C₂₄H₂₂N₂O₅
MW: 418.45 g/mol
MDL Number: MFCD02682255
CAS Number: 726181-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR