Fmoc-Tyr(3,5-I2)-OH
Supplier: Matrix Scientific
Synonyms:
Fmoc-3,5-diiodo-Tyr-OH, N-Fmoc-3,5-diiodo-L-tyrosine, Fmoc-Tyr(3,5-DiI)-OH
041942-1G
041942-5G
101853-818EA
187.69
USD
101853-818
101853-820
Fmoc-Tyr(3,5-I2)-OH
Fmoc-Tyr(3,5-I2)-OH
Hazard: Irritant
Formula:
C₂₄H₁₉I₂NO₅ MW: 655.227 g/mol |
MDL Number:
MFCD00063330 CAS Number: 103213-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...