Fmoc-3,5-diiodo-L-thyronine

Supplier: Matrix Scientific

041941-500MG 041941-1G
101853-814EA 436.92 USD
101853-814 101853-816
Fmoc-3,5-diiodo-L-thyronine
Fmoc-3,5-diiodo-L-thyronine

Hazard: Irritant

Formula: C₃₀H₂₃I₂NO₆
MW: 747.33 g/mol
MDL Number: MFCD02682265
CAS Number: 1217733-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR