Fmoc-3,5-diiodo-D-Tyr-OH

Supplier: Matrix Scientific

041940-1G 041940-5G
101853-810EA 498.46 USD
101853-810 101853-812
Fmoc-3,5-diiodo-D-Tyr-OH
Fmoc-3,5-diiodo-D-Tyr-OH

Hazard: Irritant

Formula: C₂₄H₁₉I₂NO₅
MDL Number: MFCD00270484
CAS Number: 212651-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR