Fmoc-3,4-dihydroxy-L-phenylalanine
Supplier: Matrix Scientific
041937-1G
041937-5G
101853-802EA
741.54
USD
101853-802
101853-804
Fmoc-3,4-dihydroxy-L-phenylalanine
Fmoc-3,4-dihydroxy-L-phenylalanine
Hazard: Irritant
Formula:
C₂₄H₂₁NO₆ |
MDL Number:
MFCD02682263 CAS Number: 137018-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...