Fmoc-3,4-dichloro-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-Phe(3,4-DiCl)-OH, (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,4-dichlorophenyl)propanoic acid, 3,4-Dichloro-L-phenylalanine, N-FMOC protected

041936-1G 041936-5G
101853-798EA 412.31 USD
101853-798 101853-800
Fmoc-3,4-dichloro-L-phenylalanine
Fmoc-3,4-dichloro-L-phenylalanine

Hazard: Irritant

Formula: C₂₄H₁₉Cl₂NO₄
MW: 456.33 g/mol
MDL Number: MFCD00273473
CAS Number: 177966-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR