Fmoc-3,4,5-trifluoro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,4,5-trifluorophenyl)propanoic acid, Fmoc-D-Phe(3,4,5-DiF)-OH

041933-1G 041933-5G
101853-786EA 498.46 USD
101853-786 101853-788
Fmoc-3,4,5-trifluoro-D-phenylalanine
Fmoc-3,4,5-trifluoro-D-phenylalanine

Hazard: Irritant

Formula: C₂₄H₁₈F₃NO₄
MW: 441.41 g/mol
MDL Number: MFCD00797584
CAS Number: 205526-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR