Fmoc-3-(trifluoromethyl)-D-phenylalanine
Supplier: Matrix Scientific
Synonyms:
3-(Trifluoromethyl)-D-phenylalanine, N-FMOC protected, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-(trifluoromethyl)phenyl)propanoic acid, Fmoc-D-Phe(3-CF₃)-OH
041931-1G
041931-5G
101853-778EA
1095.38
USD
101853-778
101853-780
Fmoc-3-(trifluoromethyl)-D-phenylalanine
Fmoc-3-(trifluoromethyl)-D-phenylalanine
Hazard: Irritant
Formula:
C₂₅H₂₀F₃NO₄ MW: 455.44 g/mol |
MDL Number:
MFCD00672556 CAS Number: 205526-28-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...