Fmoc-2-nitro-D-phenylalanine

Supplier: Matrix Scientific

041929-1G 041929-5G
101853-770EA 386.15 USD
101853-770 101853-772
Fmoc-2-nitro-D-phenylalanine
Fmoc-2-nitro-D-phenylalanine

Hazard: Irritant

Formula: C₂₄H₂₀N₂O₆
MW: 432.44 g/mol
MDL Number: MFCD01317714
CAS Number: 478183-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR