Fmoc-2-methyl-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: 2-Methyl-L-phenylalanine, N-FMOC protected, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(o-tolyl)propanoic acid

041928-1G 041928-5G
101853-766EA 300 USD
101853-766 101853-768
Fmoc-2-methyl-L-phenylalanine
Fmoc-2-methyl-L-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₃NO₄
MW: 401.47 g/mol
MDL Number: MFCD00671391
CAS Number: 211637-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR