Fmoc-2-cyano-D-phenylalanine

Supplier: Matrix Scientific

041925-1G 041925-5G
101853-754EA 547.69 USD
101853-754 101853-756
Fmoc-2-cyano-D-phenylalanine
Fmoc-2-cyano-D-phenylalanine

Hazard: Irritant

Formula: C₂₅H₂₀N₂O₄
MW: 412.45 g/mol
MDL Number: MFCD01863770
CAS Number: 401620-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR